Learn More
Research Products International Corp Lab-Sorb Crystals, 16 oz
SDP
Catalog No. 502229552
Encompass
Lab-Sorb converts liquid waste to solid waste in seconds for easy disposal. Super absorbent crystals rapidly absorb aqueous or mixed aqueous/organic solvent and form solid waste. Ideal for collecting liquid waste in bottom of containers and preventing leaks. One 16 ounce jar can absorb up to 50 liters of liquid waste. Supplied with durable disposal bags and scoop.
Converts liquids to solids
Cleans up spills quickly
Retains liquids during autoclaving
Catalog No. 50-222-9552 Supplier Research Products International Corp Supplier No. 249411
Lab-Sorb converts liquid waste to solid waste in seconds for easy disposal. Super absorbent crystals rapidly absorb aqueous or mixed aqueous/organic solvent and form solid waste. Ideal for collecting liquid waste in bottom of containers and preventing leaks. One 16 ounce jar can absorb up to 50 liters of liquid waste. Supplied with durable disposal bags and scoop.
Converts liquids to solids
Cleans up spills quickly
Retains liquids during autoclaving