Learn More
Accuflow Systems Saint-Gobain Chemware Lab Spoon Spatula, PTFE Coated, 225 mm
Catalog No. NC2327712
Encompass_Preferred
PTFE-coated spoon spatula with a nickel-plated stainless steel core, 225 mm overall length, designed for controlled transfer, scraping, and dispensing of powders and viscous samples while minimizing sample adhesion.
  • 225 mm (22.5 cm) overall length
  • Nickel-plated stainless steel core with PTFE fluoropolymer coating
  • Spoon (Hayman) style head for scraping and transferring small quantities
  • Heavy-duty PTFE coating reduces buildup and prevents sample adhesion
  • Resistant to low and high temperatures for chemically active materials
  • Smooth, non-scratching finish suitable for delicate surfaces
Catalog No. NC2327712 Supplier Accuflow Systems Supplier No. D1069296
PTFE-coated spoon spatula with a nickel-plated stainless steel core, 225 mm overall length, designed for controlled transfer, scraping, and dispensing of powders and viscous samples while minimizing sample adhesion.
  • 225 mm (22.5 cm) overall length
  • Nickel-plated stainless steel core with PTFE fluoropolymer coating
  • Spoon (Hayman) style head for scraping and transferring small quantities
  • Heavy-duty PTFE coating reduces buildup and prevents sample adhesion
  • Resistant to low and high temperatures for chemically active materials
  • Smooth, non-scratching finish suitable for delicate surfaces