Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex L/S Easy-Load II Pump Head with 2-Channels for Precision Pump Tubing, PPS Housing, SS Rotor
Catalog No. NC9134637
Encompass
The two-channel pump head lets you pump two synchronous flow channels, proper retention requires the use of the same size tubing in each channel. These pump heads accept the six L/S precision tubing sizes L/S 13, L/S 14, L/S 16, and L/S 25. Achieve flow rates of 0.06 to 1000 mL/min¿flow rate depends on drive rpm and tubing size.
Catalog No. NC9134637 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7720260
The two-channel pump head lets you pump two synchronous flow channels, proper retention requires the use of the same size tubing in each channel. These pump heads accept the six L/S precision tubing sizes L/S 13, L/S 14, L/S 16, and L/S 25. Achieve flow rates of 0.06 to 1000 mL/min¿flow rate depends on drive rpm and tubing size.