Learn More
Stoelting™ Lab Standard with Mouse and Neonatal Rat Adaptor
SDP
Catalog No. 10000236 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Digital Dual Standard with Adaptor
Digital Lab Standard with Adaptor
Dual Lab Standard with Adaptor
Lab Standard with Adaptor
Ultra Precise Digital Dual Standard
Ultra Precise Digital Lab Standard
Ultra Precise Dual Lab Standard
Ultra Precise Lab Standard
Animal:
Mouse
Mouse, Neonatal Rat
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Animal
10-000-236 Digital Lab Standard with Adaptor Mouse, Neonatal Rat
10-000-145 Lab Standard with Adaptor Mouse, Neonatal Rat
10-000-146 Dual Lab Standard with Adaptor Mouse, Neonatal Rat
10-000-237 Digital Dual Standard with Adaptor Mouse, Neonatal Rat
10-000-402 Ultra Precise Lab Standard Mouse
10-000-403 Ultra Precise Dual Lab Standard Mouse
10-000-424 Ultra Precise Digital Lab Standard Mouse
10-000-425 Ultra Precise Digital Dual Standard Mouse
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 10-000-236 Supplier Stoelting™ Supplier No. 51925
Add to Cart
Add to Cart
Request a Quote
Stoelting Mouse/Neonatal Rat Adaptor is designed for use with the traditional U frame Stereotaxic instrument. The adaptor employs light, Delrin Ear Bars with tapered points on one end, and rubber tips on the other, to facilitate surgery on mice and rat pups
Model organisms with soft skulls, such as the mouse and the neonatal rat, are difficult to secure firmly in a traditional stereotaxic instrument. The stainless steel ear bars used for adult rats are too heavy to provide a secure hold without compressing the sides of the skull inward (which closes the airways and impedes blood flow). Further, the neonatal rat pup has no external opening of the auditory canal into which to insert the ear bars.
  • Unique feature permits ear bars to be independently adjusted in height to level the skull
  • Laser-engraved scales show the vertical positions of the ear bars
  • Tooth Bar and Nose Clamp secure the nose
  • Jaw Holder Cuffs fit over the ends of the ear bars, replacing rubber tips, providing an alternative non-invasive means of securing the animal's head in the stereotaxic stage
  • These options for triple point securing of the animal's head in this adaptor make it usefully versatile
Specifications
For Use With (Application) Neuroscience
For Use With (Equipment) Lab Standard™ series, Stoelting, and most other Stereotaxic Instruments
Animal Mouse, Neonatal Rat
Model Digital Lab Standard with Adaptor