Learn More
Braintree Scientific Lab Standard Stereotaxic Instrument - w/18 degree ear bar
SDP
Catalog No. 501953980
Encompass
Single manipulator models read from the open end of the U. Large, easy-to-read numerals on scale. Precise alignment to 0.1mm resolution. Triple-start lead screws for fastest positioning possible. Absolute lock at 90. Universal joint allows scientist to probe in either anterior-posterior or lateral-medial planes. Manipulator. Arm may be swiveled out of the way for convenience in installing animal, or for performing a procedure, and then reliably returned to the same point. Available with 18 or 45 (non-puncture ear bars). **Sold for research purposes only and not for human use**
Catalog No. 50-195-3980 Supplier Braintree Scientific Supplier No. 51600
Single manipulator models read from the open end of the U. Large, easy-to-read numerals on scale. Precise alignment to 0.1mm resolution. Triple-start lead screws for fastest positioning possible. Absolute lock at 90. Universal joint allows scientist to probe in either anterior-posterior or lateral-medial planes. Manipulator. Arm may be swiveled out of the way for convenience in installing animal, or for performing a procedure, and then reliably returned to the same point. Available with 18 or 45 (non-puncture ear bars). **Sold for research purposes only and not for human use**