Learn More
Stoelting™ Lab Standard™ Stereotaxic Instrument for Rats
SDP
Catalog No. 10000420 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Digital Dual Lab Standard
Digital Lab Standard
Digital Lab Std,nonrupture ear bars
Dual Lab Standard
Lab Standard
Lab Standard, Base only
Lab Standard, non-rupture ear bars
Ultra Precise Digital
Ultra Precise Dual Digital
Ultra Precise Dual Lab Standard
Ultra Precise Lab Standard
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
10-000-420 Ultra Precise Dual Digital
10-000-133 Lab Standard
10-000-134 Lab Standard, Base only
10-000-136 Dual Lab Standard
10-000-230 Digital Lab Standard
10-000-231 Digital Dual Lab Standard
10-000-394 Ultra Precise Lab Standard
10-000-396 Ultra Precise Dual Lab Standard
10-000-418 Ultra Precise Digital
10-000-173 Lab Standard, non-rupture ear bars
10-000-238 Digital Lab Std,nonrupture ear bars
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 10-000-420 Supplier Stoelting™ Supplier No. 51903U
 Restricted Item
Add to Cart
Add to Cart
Request a Quote
Classic Lab Standard™ includes a 100 micron 3-axes, left-hand manipulator arm, rat adaptor, traditional (18°) ear bars, and corner clamp probe holder. Available as dual, ultra-precise, digital or motorized.
  • Large, easy to read vernier scales Scales are laser engraved — accurate to 100 microns
  • Triple lead screws for fast positioning 80 mm of vertical, lateral and anterior-posterior travel
  • Absolute lock at 90° (vertical)
  • Brass bushings in manipulator arm permit electrical grounding
  • Accessories available for use with a wide variety of small animals
  • Adaptors available for use with mice, neonatal rats, birds and other species
  • Gas anesthesia compatible
  • Easily cleaned durable surface
  • Warmer ready base compatible with Stoelting Rodent Warmer System
  • Versatile Positioning
  • Manipulator arm controls medial-lateral and ventral-dorsal using exclusive triple lead screws for faster positioning, and anterior-posterior movement via dovetail slide movement, with 80mm of travel possible in each direction.
  • Universal joint allows the researcher to change the angle of the probe up to 90° in either the anterior-posterior or mediallateral planes.
  • Improved locking mechanism on the Lab Standard™ will hold any angle position without slipping and provides an absolute lock at 90° vertical.
  • Easily-Read Scales
    • Oriented to be easily read from the open end of the U
    • Numerals are large and easy to read.
    • Scale lines are laser engraved, to allow the finest possible permanent marking of scales on all 3 axes.
    • Precise alignment with facing vernier scales gives accurate resolution to 100 microns.
  • Smooth, Consistent Movements - Triple-lead screws allow smooth consistent movements with the fastest positioning possible.
  • Warmer ready base
  • Base Plates are designed to be integrated with the Stoelting Rodent Warmer System (sold separately) and are used to maintain body temperature throughout surgical procedures.
  • Thermal heating pads are embedded into base plates to avoid cross contamination with easy clean-up and maintain a sterile environment during stereotaxic surgery.
  • Can be used as a general warming system or with a rectal thermal probe (sold separately) for homeothermic regulation.
Specifications
For Use With (Application) Neuroscience
Animal Rat
Model Ultra Precise Dual Digital