Learn More
Research Products International Corp RPI Lab Tape, 3"" Core, 1/2"" Wide, 2160"" Length, Violet
SDP
Catalog No. 501978597
Encompass
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oil proof, waterproof and acid resistant.
Catalog No. 50-197-8597 Supplier Research Products International Corp Supplier No. 561205V
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oil proof, waterproof and acid resistant.