Learn More
GA International Color Laboratory Tape 0.94" x 180' / 24mm x 55m Green Seafoam From -80°C to +121°C (-112°F to +250°F)
Catalog No. NC0213501
Encompass_Preferred
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Catalog No. NC0213501 Supplier GA International Supplier No. PAT24GS
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.