Learn More
GA International Color Laboratory Tape 0.71" x 180' / 18mm x 55m Grey From -80°C to +121°C (-112°F to +250°F)
Catalog No. NC0876042
Encompass_Preferred
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Catalog No. NC0876042 Supplier GA International Supplier No. PAT18GR
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.