Learn More
Research Products International Corp RPI Color Coded Multi-Purpose Laboratory Tape, 3 Inch Core, 1/2 Inch Wide, 2,160 Inches per Roll, Green
SDP
Catalog No. 50999323
Encompass
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oil proof, waterproof and acid resistant.
Catalog No. 50-999-323 Supplier Research Products International Corp Supplier No. 561205G
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oil proof, waterproof and acid resistant.