Learn More
GA International Color Laboratory Tape 0.94" x 180' / 24mm x 55m Orange From -80°C to +121°C (-112°F to +250°F)
Catalog No. NC0350985
Encompass_Preferred
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.
Catalog No. NC0350985 Supplier GA International Supplier No. PAT24OR
Color laboratory tape. Available in an assortment of eye-catching colors and various widths, perfect for color coding and general lab use. Can be written with permanent markers and ball-point pens for easy and quick identification of cardboard, glass, plastic, and metal surfaces.