Learn More
Research Products International Corp RPI Color Coded Multi-Purpose Laboratory Tape, 3 Inch Core, 1 Inch Wide, 2,160 Inches per Roll, Orange
SDP
Catalog No. 50998047
Encompass
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.
Catalog No. 50-998-047 Supplier Research Products International Corp Supplier No. 560105O
Laboratory tape adheres to any clean dry surface including glass, plastic, metal, paper, or rubber and removes without leaving a sticky residue. Can be marked with pencil, pen or solvent ink markers. Tape adhesive is oilproof, waterproof and acid resistant.