Learn More
Kuhner Shaker Inc LAB-THERM LT-XC WITH CO2 NO C
Catalog No. 502570037
Encompass
Lab-Therm LT-X with coolingTemperature range: ambient - 15C to 80CControl increments of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm, and User Specified)Accepts loads up to 25kg (>25kg possible if required)DC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H746 x W855 x D810mmWeight: 170kgFIVE YEAR WARRANTY HARDWARE, 1 YEAR WARRANTY ON SENSORS
Catalog No. 50-257-0037 Supplier Kuhner Shaker Inc Supplier No. SMX1701
Add to Cart
Add to Cart
Request a Quote
Lab-Therm LT-X with coolingTemperature range: ambient - 15C to 80CControl increments of 0.1C, Accuracy +/- 0.3C at 37CUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm, and User Specified)Accepts loads up to 25kg (>25kg possible if required)DC magnetic direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H746 x W855 x D810mmWeight: 170kgFIVE YEAR WARRANTY HARDWARE, 1 YEAR WARRANTY ON SENSORS