Learn More
Kuhner Shaker Inc LAB-THERM LT-XC WITH COOLING A
Catalog No. 502570036
Encompass
LT-X with humidity, EcoDewTemperature range: ambient + 10C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CHumidity control 75-85%+/-0.3%RH @<22CRTUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H841 x W855 x D810mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS
Catalog No. 50-257-0036 Supplier Kuhner Shaker Inc Supplier No. SMX1702
Add to Cart
Add to Cart
Request a Quote
LT-X with humidity, EcoDewTemperature range: ambient + 10C to 80CControl of 0.1C, Accuracy +/- 0.3C at 37CHumidity control 75-85%+/-0.3%RH @<22CRTUser Selectable Orbit Diameter (12.5mm, 25mm, 50mm)Loads up to 25kg (+25kg possible if required)DC direct drive with single eccentric (no belt)100% stainless steel interiorExternal Dimensions: H841 x W855 x D810mmWeight: 210kgFIVE YEAR WARRANTY HARDWARE,1 YEAR WARRANTY ON SENSORS