Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Berkshire™ Lab-Tips™ Low TOC Swab Large

Catalog No. 19066704 Shop All Berkshire Corporation Products
Change view
Click to view available options
Length (Metric):
100 mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
19-066-704 100 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-066-704 Supplier Berkshire™ Supplier No. LTP125TOC.25

Berkshire Lab-Tips™ Low TOC Swab Large knitted polyester swab features a unique semi-flexible internal paddle for greater surface coverage.

  • Specifically designed for Total Organic Carbon analysis and TOC tests in low-TOC environment
  • Critically low particles, fibers, ions and extractables
  • Excellent chemical resistance
  • High abrasion resistance while soft and non-abrasive
  • No contaminating adhesives and good sorbency

Specifications

Quantity 20 swabs/pack; 25 packs/case
Length (English) 3.94 in.
Product Type TOC Swab
Length (Metric) 100 mm
Material Polyester
Size Large
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.