Learn More
Accuflow Systems Saint-Gobain Chemware Lab Tongs, PTFE Coated, Stainless Steel, 240 mm
Catalog No. NC2327709
Encompass_Preferred
Saint-Gobain Chemware PTFE Coated Lab Tongs are 240 mm stainless steel tongs fully covered with an extra-heavy PTFE coating to provide a non-stick, chemically resistant tool for handling, dipping, and manipulating delicate or corrosive samples in the laboratory.
  • PTFE-coated stainless steel construction provides a non-stick surface.
  • 240 mm (9.38 in) length suited for general lab handling and dipping tasks.
  • Protects delicate surfaces and minimizes sample loss through sticking.
  • Suitable for use with corrosive media when the coating remains intact.
  • Maximum temperature 288°C (550°F); not recommended for cryogenic use.
  • Six month manufacturer warranty.
Catalog No. NC2327709 Supplier Accuflow Systems Supplier No. D1069290
Saint-Gobain Chemware PTFE Coated Lab Tongs are 240 mm stainless steel tongs fully covered with an extra-heavy PTFE coating to provide a non-stick, chemically resistant tool for handling, dipping, and manipulating delicate or corrosive samples in the laboratory.
  • PTFE-coated stainless steel construction provides a non-stick surface.
  • 240 mm (9.38 in) length suited for general lab handling and dipping tasks.
  • Protects delicate surfaces and minimizes sample loss through sticking.
  • Suitable for use with corrosive media when the coating remains intact.
  • Maximum temperature 288°C (550°F); not recommended for cryogenic use.
  • Six month manufacturer warranty.