Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Transfer Tubing, DEHP-Free Flexible PVC, 3/32"" ID x 5/32"" OD; 50 ft
Catalog No. NC1681093
Encompass
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications. Characteristics:Nontoxic DEHP-free formulation. Flexible, small bend radii. Transparent; allows for visual inspection and flow monitoring. Certifications: FDA compliant, Proposition 65, REACH Temperature range: -58 to 165¿F (-50 to 73¿C) Sterilize: Autoclave.
Catalog No. NC1681093 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX9550502
Applications: Analytical instruments, general laboratory. Use for transfer of liquids, gases, and inorganic chemicals found in lab, paint, pigment, and soap dispensing applications. Characteristics:Nontoxic DEHP-free formulation. Flexible, small bend radii. Transparent; allows for visual inspection and flow monitoring. Certifications: FDA compliant, Proposition 65, REACH Temperature range: -58 to 165¿F (-50 to 73¿C) Sterilize: Autoclave.