Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Antibody Diluent OP Quanto

Catalog No. TA125ADQ Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA125ADQ 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TA125ADQ Supplier Epredia™ Supplier No. TA125ADQ
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Achieve crisp staining while minimizing background staining with Epredia™ Lab Vision™ Antibody Diluent OP Quanto.

  • Contains a protein stabilizing agent and preservative
  • Suitable for use as a negative control reagent during immunohistochemical staining
  • Concentrated antibodies can be diluted to the appropriate titer specified on the individual antibody datasheet or determined by the user, to achieve optimal staining

Specifications

Capacity 125 mL
Content And Storage 2° to 8°C
Product Type Antibody Diluent
Quantity 125 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.