Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Autostainer 360-2D, Autostainer 360 and PT Module 110V

Catalog No. p-6024368 Shop All Epredia Products
Click to view available options
Product Type:
Autostainer 360 + PT Module
Voltage:
110 V
Products on this page may include voltages not considered standard in North America. Please review carefully.
Products on this page may include voltages not considered standard in North America. Please review carefully.

Stain at least 108 slides per day with this fast, space-saving unit that provides flexible options to meet the workflow needs of any laboratory.

Benefits:
  • One to 36 slide capacity, each individually programmable
  • User-specified protocols and choice of reagents
  • Multiple staining runs per day
  • Truly hands-free, walk-away operation with advanced timer functions for overnight use
  • Easy, Windows™ 7 based programming
  • Modular options with up to four units controlled from one PC
  • Complemented by the Epredia™ PT Module designed for fully automated, simultaneous dewaxing and epitope recovery (The PT Module is available separately or in a system package).
  • Fast flow software logic enables flexible programming, operator ease of use and control of IHC costs

System Features:

  • Incorporates 2D Data Matrix Bar Coding for Slides and Reagents
  • Can be configured for research or diagnostic applications at no additional cost
  • Scans rapidly using high precision label detector technology 
  • Has integrated digital bar code scanner
  • Fully configured 2D data matrix software and printer interface
  • Bar code control of all protocol, reagent and slide operations
  • Preprintable data matrix labels with user-defined autoprograms
  • Adaptable for proprietary 2D-labeled reagents or user-defined reagents/labels
  • Optional individual inactivation of slide and/or reagent bar coding

Includes:

  • 36-slide Autostainer with 2D matrix bar code reader
  • Computer
  • Monitor
  • Label printer(s)
  • Document printer
  • Three metal reinforced, ABS plastic slide racks
  • Reagent rack Reagent vials and caps
  • Autostainer probe cleaning reagent kit
  • Buffer rinse bottle
  • Wash Buffer, DI water and waste containers
  • Universal Power Supply

Specifications

Product Type Autostainer 360 + PT Module
Slide Capacity 1 to 36 Slides/Run
Carryover <10-6
Reagent Capacity Up to 40 reagents/run, 12mL/reagent
Temperature Range (Metric) Ambient
Weight (Metric) ∼54 kg
Height (Metric) 58 cm
Length (Metric) 67 cm
Width (Metric) 76 cm
Voltage 110 V
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.