Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ biotinylated Goat anti-Polyvalent (Ready-To-Use)

Catalog No. TP125BN Shop All Epredia Products
Change view
Click to view available options
Quantity:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TP125BN 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. TP125BN Supplier Epredia™ Supplier No. TP125BN
This item is not returnable. View return policy

Employ a labeled streptavidin-biotin immunoenzymatic antigen detection system with the Epredia™ Lab Vision™ Biotinylated Goat anti-Polyvalent (Ready-To-Use).

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate-chromogen.
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Conjugate: Biotin

Specifications

Conjugate Biotin
Content And Storage 2° to 8°C
Product Type Anti-Polyvalent
Quantity 125mL
Species anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.