Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X)

Catalog No. AP9003500 Shop All Epredia Products
Change view
Click to view available options
Quantity:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP9003500 500mL
AP9003125 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AP9003500 Supplier Epredia™ Supplier No. AP9003500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve the reactivity of many antibodies in formalin-fixed tissues with the Epredia™ Lab Vision™ Citrate Buffer for Heat-Induced Epitope Retrieval (10X).

  • Citrate buffer, pH 6.0
  • 10X stock solution
  • Diluted 10-fold with distilled water before use
Storage Conditions:
  • Store at room temperature.
  • Stable for up to 18 months.
  • Contains no preservatives.
  • For periods longer than 3 months, store at 2-8°C.

Specifications

Content And Storage Room temperature
Dilution 10X
Product Type Citrate Buffer
Quantity 500mL
For Use With (Application) Immunohistochemistry
pH 6

For Reserach Use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.