Learn More
Description
- 10mM EDTA buffer, pH 8.0
- Dilute 10-fold with distilled water before use
Storage Conditions: Store at room temperature. Stable for up to 18 months. Contains no preservatives. For periods longer than 3 months, store at 2-8°C
Specifications
Specifications
| Content And Storage | Room temperature |
| Dilution | 10X |
| Product Type | EDTA |
| Quantity | 125mL |
| For Use With (Application) | Immunohistochemistry |
| pH | 8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.