Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ HIER Buffer L (100X)

Catalog No. TA135HBL Shop All Epredia Products
Change view
Click to view available options
Quantity:
9 x 15 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA135HBL 9 x 15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TA135HBL Supplier Epredia™ Supplier No. TA135HBL
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Perform heat induced epitope retrieval (HIER) on formalin-fixed paraffin-embedded tissue sections using the Epredia™ Lab Vision™ HIER Buffer L (100X).

  • For in vitro diagnostic use
  • Storage Conditions: Store at room temperature

Specifications

Content And Storage Room temperature
Dilution 100X
Product Type HIER
Quantity 9 x 15 mL
For Use With (Application) Immunohistochemistry
pH 6 to 7
Regulatory Status IVD
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.