Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Mayer's Hematoxylin

Catalog No. ta060mh Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA060MH 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TA060MH Supplier Epredia™ Supplier No. TA060MH

Designed for use in histological applications, Epredia™ Lab Vision™ Mayer's Hematoxylin provides superior quality.

  • Requires slightly longer application periods
  • Based on the original formulation developed by Mayer

Specifications

Content And Storage Room temperature
Product Type Hematoxylin
Quantity 60 mL
For Use With (Application) Healthcare

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.