Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval

Catalog No. AP9007005 Shop All Epredia Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP9007005 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP9007005 Supplier Epredia™ Supplier No. AP9007005

missing translation for 'validMainframeMsg'

Improve the immunoreactivity of many antigens in formaldehyde fixed tissues with Epredia™ Lab Vision™ Pepsin Solution for Enzyme-Induced Epitope Retrieval.

  • pH 2.0
  • Single-component, ready-to-use format

Storage Conditions: Store at 4°C. Stable for 12 months when stored at 2° to 8°C.

Specifications

Capacity 5 mL
Content And Storage Store at 4°C
Product Type Pepsin
Quantity 5 mL
For Use With (Application) Immunohistochemistry
pH 2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.