Learn More
Epredia™ Lab Vision™ Permanent Fast-Red Quanto
Catalog No. TA125QAL Shop All Epredia Products
Change view
Click to view available options
125mL (415-625 tests)
Catalog No.
TA125QAL 125mL (415-625 tests)
1 options
Catalog No. TA125QAL Supplier Epredia™ Supplier No. TA125QAL
Epredia™ Lab Vision Permanent Fast-Red Quanto provides a permanent magenta stain for use with immunohistochemistry procedures.
Historically, integrating fast red chromogens into routine Imunohistochemistry was a challenge. Epredia Permanent Fast-Red Quanto eliminates these challenges and produces the brilliant results you expect.
  • Permanent Results — Does not fade during long term storage
  • Solvent Resistance — Insoluable in xylene or alcohols allowing Fast-Red slides to be coverslipped with routine HEs and IHC slides
  • Automated “On Slide” Chromogen Mixing — The two chromogen reagents are delivered in equal parts and are mixed sequentially, directly on the slide, allowing any open instrument to mix the chromogen on board reducing turn around time and manual steps
  • Fast Incubation Times — With typical incubation times of 11 to 15 minutes Permanent Fast Red Quanto is one of the quickest fast reds on the market
Specifications
Substrate Quantity 125mL