Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval

Catalog No. AP9006005 Shop All Epredia Products
Change view
Click to view available options
Quantity:
2mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP9006005 5mL
AP9006002 2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AP9006005 Supplier Epredia™ Supplier No. AP9006005
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve the immunoreactivity of certain antigens in formaldehyde-fixed tissues with Epredia™ Lab Vision™ Protease XXV for Enzyme-Induced Epitope Retrieval.

Storage Conditions: Store at 2°-8°C. Stable for 18 months

Specifications

Content And Storage 2° to 8°C.
Concentration 1 mg/mL
Product Type Protease
Quantity 5mL
For Use With (Application) Immunohistochemistry
pH 7.4
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.