Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ PT Module
GSA_VA

Catalog No. a80400012 Shop All Epredia Products
Change view
Click to view available options
Slide Capacity:
48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Slide Capacity
A80400012 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A80400012 Supplier Epredia™ Supplier No. A80400012
Only null left
Add to Cart
Add to Cart
Request a Quote

Automatic pre-treatment module standardizes the manual steps associated with the pre-treatment of slides. Simultaneously perform dewaxing and antigen retrieval on slides prior to immunohistochemical staining.

Module offers consistent antigen retrieval for high quality immunohistochemistry. Using gentle heat, the retrieval device quickly produces outstanding results while maintaining the integrity of delicate tissue. In addition, most users can streamline testing practices by consolidating various antigen retrieval methods into only one or two PT Module antigen retrieval device procedures that can be run simultaneously. Precise temperature control without the damaging effects of temperature variations, microwaves or pressure delivers a level of consistency unmatched by any other instrument available.

  • Easy to use, one-touch operation via built-in touchscreen
  • Operator defined, preprogrammed time and temperature settings
  • Dual reaction baths, each with 24-slide capacity, for simultaneous operation of two independent procedures
  • Proprietary epitope recovery solutions for safe, nontoxic, solvent-free and environmentally improved dewaxing
  • Provides for rapid, gentle heating, eliminating microwave and pressure cooker variability
  • Maintains excellent tissue/cellular morphology
  • Efficient, time-saving transfer of fully compatible slide racks directly to any Autostainer after treatment to avoid slide manipulation
  • Fully programmable for overnight operation or delayed-start, early-morning pre-heating
  • Can be connected to a remote computer to monitor and log antigen retrieval reports for regulatory compliance.
  • The PT Module is fully automated and includes PT Monitor software for printing/maintenance of regulatory compliant procedural reports.
Note: This item has US voltage and is not suitable for use in UK.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Slide Capacity 48
Reagent Capacity 1.5 L
Temperature Range (Metric) 5°C to 40°C
Amperage 10 A max 120 VAC
Frequency 50/60 Hz
Voltage 100/120 V
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.