Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lab Vision™ Tris-Buffered Saline (25X), Epredia™

Catalog No. TA125TB Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TA125TB 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TA125TB Supplier Epredia Supplier No. TA125TB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Take advantage of a wash buffer for immunolabeling techniques with the Epredia™ Lab Vision™ Tris-Buffered Saline (25X),

Storage Conditions: Store at room temperature. Stable for 2 years from date of manufacture. Use diluted solution within one week.

Specifications

Chemical Name or Material Tris-Buffered Saline (25X)
Concentration 25X
For Use With (Application) Healthcare
Quantity 125 mL

For in vitro diagnostic use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.