Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X)

Catalog No. AP9005050 Shop All Epredia Products
Change view
Click to view available options
Quantity:
50mL
125mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AP9005050 50mL
AP9005125 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AP9005050 Supplier Epredia™ Supplier No. AP9005050
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Improve the reactivity of certain antibodies in formalin-fixed tissues with Epredia™ Lab Vision™ Tris-HCl Buffer for Heat-Induced Epitope Retrieval (10X).

  • 100X stock solution
  • 100mM Tris-HCl buffer, pH 10.0
  • Dilute 10-fold with distilled water before use

Storage Conditions: Store at 2-8°C. Stable for up to 18 months. Contains no preservatives.

Specifications

Content And Storage 2° to 8°C.
Dilution 10X
Product Type HIER
Quantity 50mL
For Use With (Application) Immunohistochemistry
pH 10
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.