Learn More
Epredia™ Lab Vision™ Tween™ 20 Detergent
Catalog No. TA125TW Shop All Epredia Products
Change view
Click to view available options
Volume (Metric):
125 mL
Catalog No. Volume (Metric)
TA125TW 125 mL
1 options
Catalog No. TA125TW Supplier Epredia™ Supplier No. TA125TW
Add to Cart
Add to Cart
Decrease background staining and enhances reagent spreading in automated and manual procedures with Epredia™ Lab Vision™ Tween™ 20 detergent.
Recommended for:
Immunohistochemistry.
Specifications
Product Line Tween 20
Content And Storage Store at Room Temperature
Volume (Metric) 125 mL