Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ UltraVision™ Detection System: anti-Polyvalent, AP (Ready-To-Use)

Catalog No. tp125al Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
125 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TP125AL 125 mL
TP060AL 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. TP125AL Supplier Epredia™ Supplier No. TP125AL

missing translation for 'validMainframeMsg'

A versatile and robust biotin-streptavidin detection system for research applications and cost-conscious laboratories.

This technique involves the sequential incubation of the specimen with an unconjugated primary antibody specific to the target antigen, a biotinylated secondary antibody that reacts with the primary antibody, enzyme-labeled streptavidin, and substrate/chromogen.
  • Labeled streptavidin-biotin immunoenzymatic antigen detection system
  • Species: Goat

  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Alkaline Phosphatase

  • Chromogen/Substrate: None provided

Includes:

TP-060-AL: 60mL Ultra V Block (TA-060-UB), 60mL Biotinylated Goat Anti-Polyvalent (TP-060-BN), 60mL Streptavidin Alkaline Phosphatase (TS-060-AP)

TP-125-AL: 125mL Ultra V Block (TA-125-UB), 125mL Biotinylated Goat Anti-Polyvalent (TP-125-BN), 125mL Streptavidin Alkaline Phosphatase (TS-125-AP)

Note:

UltraVision detection system detects a specific antibody bound to an antigen in tissue sections. The specific antibody is located by a biotin-conjugated secondary antibody. This step is followed by the addition of a streptavidin-enzyme conjugate that binds to the biotin present on the secondary antibody. The specific antibody, secondary antibody, and streptavidin-enzyme complex is then visualized with an appropriate substrate/chromogen.

Specifications

Chromagen None provided
Count 625-1250 slides
Enzyme Alkaline Phosphatase
Product Line Lab Vision, UltraVision
Species Goat
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Substrate None provided
Includes 125mL Ultra V Block (TA-125-UB), 125mL Biotinylated Goat Anti-Polyvalent (TP-125-BN), 125mL Streptavidin Alkaline Phosphatase (TS-125-AP)
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.