Learn More
Description
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Enzyme: Peroxidase
Chromogen/Substrate: Diaminobenzidine (DAB)
Includes:
TL-015-HD: 15mL Hydrogen Peroxide Block (TA-015-HP), 15mL Ultra V Block (TA-015-UB), 15mL Primary Antibody Enhancer (TL-015-PB), 15mL HRP Polymer (TL-015-PH), 15mL DAB Plus Substrate (TA-015-HSX), 1mL DAB Plus Chromogen (TA-001-HCX)
TL-060-HD: 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX)
TL-125-HD: 125mL Hydrogen Peroxide Block (TA-125-HP), 125mL Ultra V Block (TA-125-UB), 125mL Primary Antibody Enhancer (TL-125-PB), 125mL HRP Polymer (TL-125-PH), 125mL DAB Plus Substrate (TA-125-HSX), 4mL DAB Plus Chromogen (TA-004-HCX)
Note:
UltraVision detection system detects a specific mouse IgG or rabbit IgG antibody bound to an antigen in tissue sections. The specific antibody is located by a universal secondary antibody formulation conjugated to an enzyme-labeled polymer that recognizes mouse and rabbit immunoglobulins. The polymer complex is then visualized with an appropriate substrate/chromogen.
Specifications
Specifications
| Quantity | 60 mL |
| Count | 300 to 600 slides |
| Includes | 60mL Hydrogen Peroxide Block (TA-060-HP), 60mL Ultra V Block (TA-060-UB), 60mL Primary Antibody Enhancer (TL-060-PB), 60mL HRP Polymer (TL-060-PH), 60mL DAB Plus Substrate (TA-060-HSX), 2mL DAB Plus Chromogen (TA-002-HCX) |
| Chromagen | Diaminobenzidine (DAB) |
| Specificity | anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L) |
| Storage Requirements | Store at 2° to 8°C. Each component is stable for 18 months. |
| Enzyme | Peroxidase |
| Substrate | Diaminobenzidine (DAB) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.