Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ UltraVision™ ONE Detection System: AP Polymer (Ready-To-Use)

Catalog No. TL060ALJ Shop All Epredia Products
Change view
Click to view available options
Quantity:
60 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL060ALJ 60 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL060ALJ Supplier Epredia™ Supplier No. TL060ALJ

missing translation for 'validMainframeMsg'

A one-step polymer system providing increased sensitivity, time savings and detection simplicity.

UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE AP polymer is an innovative, patented technology consisting of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE AP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This biotin-free system eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Alkaline Phosphatase

  • Chromogen/Substrate: None provided

Includes:

TL-060-ALJ: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE AP Polymer (TL-060-APJ)

TL-125-ALJ: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE AP Polymer (TL-125-APJ)

Note:

UltraVision ONE AP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to alkaline phosphatase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.

Specifications

Chromagen None provided
Count 300 to 600 slides
Enzyme Alkaline Phosphatase
Product Line Lab Vision, UltraVision
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Substrate None provided
Includes 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE AP Polymer (TL-060-APJ)
Quantity 60 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.