Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ UltraVision™ ONE Detection System: HRP Polymer (Ready-To-Use)

Catalog No. TL125HLJ Shop All Epredia Products
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
TL125HLJ 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. TL125HLJ Supplier Epredia™ Supplier No. TL125HLJ

missing translation for 'validMainframeMsg'

A one-step polymer system providing increased sensitivity, time savings and detection simplicity.

UltraVision ONE is a robust ONE-step polymer system that provides increased sensitivity, time savings and detection simplicity. The UltraVision ONE HRP polymer is an innovative, patented technology. It consists of smaller amino acid polymer subunits that minimize conflicts in binding the target protein. Decreased binding conflicts result in more consistent staining and better signal amplification. Ultimately, this gives the user higher sensitivity and antibody efficiency. UltraVision ONE HRP polymer allows the use of less antibody to obtain better signal-to-noise ratios. This system is also biotin-free, which eliminates background staining found with traditional biotin-based detection methods. For optimal interpretation of results, appropriate positive and negative controls must be included.
  • Specificity: anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)

  • Enzyme: Peroxidase

  • Chromogen/Substrate: None provided

Includes:

TL-060-HL: 60mL Ultra V Block (TA-060-UB), 60mL UltraVision ONE HRP Polymer (TL-060-PHJ)

TL-125-HL: 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)

Note:

UltraVision ONE HRP polymer detection system detects rabbit and mouse immunoglobulins bound to an antigen in tissue sections. The specific primary antibody is located by a universal secondary antibody polymer formulation. The amino acid polymer is conjugated to horseradish peroxidase and the Fab fragments of goat anti-rabbit and goat anti-mouse. The polymer complex is then visualized with an appropriate substrate/chromogen.

Specifications

Quantity 125 mL
Count 625 to 1250 tests
Includes 125mL Ultra V Block (TA-125-UB), 125mL UltraVision ONE HRP Polymer (TL-125-PHJ)
Chromagen None provided
Specificity anti-Mouse IgG (H+L), anti-Rabbit IgG (H+L)
Storage Requirements Store at 2° to 8°C. Each component is stable for 18 months.
Enzyme Peroxidase
Product Line Lab Vision, UltraVision
Substrate None provided
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.