Learn More
Alkali Scientific Lab Wash Bottles – Wide Mouth, Jet Tip, PP Closure and PPCO Draw Tube, LDPE Material, Chemical Resistant, 250mL, 6/Pk
SDP
Catalog No. 5000662473
Encompass_Preferred
Crafted for dependable daily use, our Plain Wash Bottles are constructed from premium low density polyethylene (LDPE), offering excellent flexibility, chemical resistance, and long lasting durability. Each bottle includes a one piece polypropylene (PP) closure that maintains its shape even after repeated use, ensuring a tight and reliable seal. The square shouldered design and wide mouth opening allow quick, mess free filling, while the precision jet tip enables fine, controlled dispensing of solvents, water, and other laboratory reagents. Ideal for rinsing glassware or dispensing cleaning solutions, these bottles deliver consistent performance across many applications. Features include durable LDPE construction for flexibility and resistance, secure one piece PP closures, wide mouth and square shoulders for easy filling, and a precision jet tip for accurate dispensing in laboratory, industrial, and educational settings.
Catalog No. 50-006-62473 Supplier Alkali Scientific Supplier No. WB0250
Crafted for dependable daily use, our Plain Wash Bottles are constructed from premium low density polyethylene (LDPE), offering excellent flexibility, chemical resistance, and long lasting durability. Each bottle includes a one piece polypropylene (PP) closure that maintains its shape even after repeated use, ensuring a tight and reliable seal. The square shouldered design and wide mouth opening allow quick, mess free filling, while the precision jet tip enables fine, controlled dispensing of solvents, water, and other laboratory reagents. Ideal for rinsing glassware or dispensing cleaning solutions, these bottles deliver consistent performance across many applications. Features include durable LDPE construction for flexibility and resistance, secure one piece PP closures, wide mouth and square shoulders for easy filling, and a precision jet tip for accurate dispensing in laboratory, industrial, and educational settings.