Learn More
Research Products International Corp Cleanware Laboratory Waste Bags, 8 x 10 Inches, 50 per Case
SDP
Catalog No. 50998759
Encompass
Disposable, white waste bags free bench or hood space when hung from walls, cabinets or virtually anywhere. Peel backing off the adhesive strip across the top and hang close to your work area. Back panel is longer than the front for easy opening. High Molecular Weight High Density (HMHD) polyethylene and steam autoclavable at 121˚C (250˚F). Maximum weight of contents should not exceed 1.36kg (3 lb).
Catalog No. 50-998-759 Supplier Research Products International Corp Supplier No. F131741008
Disposable, white waste bags free bench or hood space when hung from walls, cabinets or virtually anywhere. Peel backing off the adhesive strip across the top and hang close to your work area. Back panel is longer than the front for easy opening. High Molecular Weight High Density (HMHD) polyethylene and steam autoclavable at 121˚C (250˚F). Maximum weight of contents should not exceed 1.36kg (3 lb).