Learn More
Thermco Products Inc Laboratory Waterproof HACCP Tested with Probe Digital Thermometer
Catalog No. NC1405638
Encompass_Preferred
NIST Traceable w/Certificate Waterproof, HACCP Tested, 4”x.125” Insertion Probe & Cable, Minimum / Maximum - Hold Functions, Programmable High/Low Alarm, Magnet and Bench Stand, C/F Switchable, Range: -40º to 200ºC (-40°to 400°F), Resolution: 0.1º, Accuracy: ± 1.0ºC between -25° to 150° 1.0° remaining range
Catalog No. NC1405638 Supplier Thermco Products Inc Supplier No. ACCD101DIG
NIST Traceable w/Certificate Waterproof, HACCP Tested, 4”x.125” Insertion Probe & Cable, Minimum / Maximum - Hold Functions, Programmable High/Low Alarm, Magnet and Bench Stand, C/F Switchable, Range: -40º to 200ºC (-40°to 400°F), Resolution: 0.1º, Accuracy: ± 1.0ºC between -25° to 150° 1.0° remaining range