Learn More
Rice Lake Weighing System Weight Set,100g-1g ASTM Class 1 SST Polished (5321) with NVLAP Accredited NIST Traceable Calibration Report
Catalog No. NC3627202
Encompass_Preferred
100 g Range, Calibration Weight Set, ASTM Class 1Stainless steel polished finishDesigned to meet ASTM E617-23 specificationsASTM Class 0 weight sets and individual weights are not individually marked with their weight value or individually serialized per ASTM E617-23Nominal value marked on weightsSerial numbers applied to case (weights may be marked with symbol)Density 7.95g/cm3
Catalog No. NC3627202 Supplier Rice Lake Weighing System Supplier No. 12461TC
100 g Range, Calibration Weight Set, ASTM Class 1Stainless steel polished finishDesigned to meet ASTM E617-23 specificationsASTM Class 0 weight sets and individual weights are not individually marked with their weight value or individually serialized per ASTM E617-23Nominal value marked on weightsSerial numbers applied to case (weights may be marked with symbol)Density 7.95g/cm3