Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Lab Wipes PROMO
SDP

Catalog No. 22026345PM Shop All Fisherbrand Products
Change view
Click to view available options
Dimensions (L x W):
12 x 13 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-026-345PM 12 x 13 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-026-345PM
This item is not returnable. View return policy

Highly absorbent and low-lint general purpose wipe for use throughout the clinical and research lab setting

  • As an alternative to paper towels and gauze, these wipes are ideal for pathology grossing and cover slipping, as well as general lab cleaning.
  • Designed for use in labs for general cleaning, small spills, and hazardous spill containment
  • Large size: 12 in. x 13 in.
  • Heavy-duty, cloth-like material is low-lint, no loose fibers or threads
  • Available in 50 Pk/Bag, 18 Bag/Cs. (900/Cs.)

Specifications

Product Type Science Precision Wipes
Length (English) 12 in.
Length (Metric) 10.97 cm
Width (English) 13 in.
Width (Metric) 11.88 cm
Material Cloth
Color White
Dimensions (L x W) 12 x 13 in.
For Use With (Application) General cleaning, small spills, and hazardous spill containment
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.