Learn More
Uline Lab Workbench without Bottom Shelf - 72 x 36", Phenolic Top
Catalog No. NC2821452
Encompass
Stands up to corrosive chemicals. Recommended for labs, chemical plants and science classrooms.Phenolic top resists heat up to 350°F. Non-porous, scratch and impact resistant.Withstands harsh acids, moisture and bacteria.Heavy-gauge steel frame with sturdy square tube legs.Adjustable height from 29 to 39" in 2" increments.
Catalog No. NC2821452 Supplier Uline Supplier No. H10528
Stands up to corrosive chemicals. Recommended for labs, chemical plants and science classrooms.Phenolic top resists heat up to 350°F. Non-porous, scratch and impact resistant.Withstands harsh acids, moisture and bacteria.Heavy-gauge steel frame with sturdy square tube legs.Adjustable height from 29 to 39" in 2" increments.