Learn More
Electron Microscopy Sciences Kimball Physics LaB6 Cathode for Leica, Cambridge
SDP
Catalog No. 5026875
Encompass
LaB6 Cathodes are high performance, resistively heated, thermionic electron source for use in SEM, TEM, electronlithography systems, and x-ray sources. They offer an extended life with thousands of hours in clean vacuum, exceptional thermal, chemical, and electrical stability, high brightness/low energy spread with oriented single crystal made from the best quality/high purity material. Style 90-15 : Standard Microflat
Catalog No. 50-268-75 Supplier Electron Microscopy Sciences Supplier No. 8108020
LaB6 Cathodes are high performance, resistively heated, thermionic electron source for use in SEM, TEM, electronlithography systems, and x-ray sources. They offer an extended life with thousands of hours in clean vacuum, exceptional thermal, chemical, and electrical stability, high brightness/low energy spread with oriented single crystal made from the best quality/high purity material. Style 90-15 : Standard Microflat