Learn More
LabAid Isolation Gown
Catalog No. 22363008
Change view
Click to view available options
Size:
Universal
Catalog No. Size
22-363-008 Universal
1 options
This item is not returnable. View return policy
Catalog No. 22-363-008 Supplier Biomed Resources Inc Supplier No. LA051600
Add to Cart
Add to Cart
This item is not returnable. View return policy
LabAid isolation gowns are ideal for animal rooms, tissue culture rooms, clinical labs and hospital settings.
Specifications
Size Universal
Gender Unisex
No. of Pockets None
Product Type General Purpose Gown
Material Polyethylene
Color Blue
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Seam Type Perforated
Autoclavable Not Autoclavable