Learn More
FUJIFILM BIOSCIENCES INC LabAssay NEFA Kit, 500 tests, 4 μL sample volume, 0.4-1.97 mEq/L assay range, 2-10°C storage
Catalog No. NC3354950
Encompass_Preferred
The LabAssay NEFA Kit is an enzymatic colorimetric assay designed for the quantitative determination of non-esterified fatty acids (NEFA). This kit supports research on lipid metabolism and metabolic disorders, providing a precise tool for measuring NEFA concentrations in biological samples.
  • Designed for non-esterified fatty acids determination
  • Utilizes enzymatic colorimetric detection method
  • Suitable for various animal serum/plasma and culture supernatants
  • Compatible with samples from mice, rats, humans, cats, and dogs
  • Fast reaction time of approximately 10 min
  • Provides 500 tests per kit
  • Storage condition: 2-10 °C
  • Research use only, not for diagnostic procedures
  • Includes reagents and standard solution for accurate results
Catalog No. NC3354950 Supplier FUJIFILM BIOSCIENCES INC Supplier No. 29994301
Add to Cart
Add to Cart
The LabAssay NEFA Kit is an enzymatic colorimetric assay designed for the quantitative determination of non-esterified fatty acids (NEFA). This kit supports research on lipid metabolism and metabolic disorders, providing a precise tool for measuring NEFA concentrations in biological samples.
  • Designed for non-esterified fatty acids determination
  • Utilizes enzymatic colorimetric detection method
  • Suitable for various animal serum/plasma and culture supernatants
  • Compatible with samples from mice, rats, humans, cats, and dogs
  • Fast reaction time of approximately 10 min
  • Provides 500 tests per kit
  • Storage condition: 2-10 °C
  • Research use only, not for diagnostic procedures
  • Includes reagents and standard solution for accurate results