Learn More
FUJIFILM Biosciences LabAssay™ Triglyceride, 350 tests, Enzymatic Colorimetric (GPO-DAOS method), 2-10 °C storage, Animal serum/plasma, 37 °C incubation, 2 μL sample volume, 600 nm wavelength measurement, 100-888 mg/dL standard curve range
Catalog No. NC2044372
Encompass_Preferred
LabAssay™ Triglyceride, 350 tests, Enzymatic Colorimetric (GPO-DAOS method), 2-10 °C storage, Animal serum/plasma, 37 °C incubation, 2 μL sample volume, 600 nm wavelength measurement, 100-888 mg/dL standard curve range
Catalog No. NC2044372 Supplier FUJIFILM Biosciences Supplier No. 29194501
LabAssay™ Triglyceride, 350 tests, Enzymatic Colorimetric (GPO-DAOS method), 2-10 °C storage, Animal serum/plasma, 37 °C incubation, 2 μL sample volume, 600 nm wavelength measurement, 100-888 mg/dL standard curve range