Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Genesis BPS™ LABath Clean

Catalog No. 22050481 Shop All Globe Scientific Products
Change view
Click to view available options
Quantity:
240 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-050-481 240 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-050-481 Supplier Genesis BPS™ Supplier No. BPS428SE530
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory and blood bank cleaning solution formulated for maintaining thawing baths, water baths, and related processing equipment

  • Designed for routine cleaning procedures where consistency and hygiene matter
  • Easy-to-use solution helps teams maintain equipment as part of daily or scheduled protocols
  • Supports blood bank, plasma thawing, and laboratory processing environments
  • Useful accessory for facilities operating thawing baths or similar laboratory equipment

Specifications

Product Type Cleaning Solution
Color-coded No
Form Concentrate
Scent Unscented
Sterility Non-sterile
Container Type Bottle
Disposable Single-use
Flammable No
For Use With (Equipment) Water Bath
Quantity 240 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.