Learn More
Labcon North America Eclipse FlexTop 1250L Extended Length Pipet Tips with UltraFine points, in Eclipse Refills, 4800/case
Catalog No. NC1836069
Encompass_Preferred
Labcon Eclipse™ FlexTop™ Low Force pipet tips are designed to fit onto most popular pipettors and eject with forces shown to help reduce the risk of strain injury. By using these Low Force tips you don¿t have to invest in special pipettors to gain all the advantages of low force pipetting. These ultimate tips feature a uniquely thin and flexible top that conforms to the pipet barrel unlike any other tips. They also include a non-beveled point that is so precise you can easily expel the tiniest amount of sample. And the top mounting is beveled to ensure that multichannel pipettes are easy to mount. FlexTop™ tip are made of clear 100% virgin polypropylene. Like all Labcon tips these tips are autoclavable along with their racks.
Catalog No. NC1836069 Supplier Labcon North America Supplier No. 10452600009
Labcon Eclipse™ FlexTop™ Low Force pipet tips are designed to fit onto most popular pipettors and eject with forces shown to help reduce the risk of strain injury. By using these Low Force tips you don¿t have to invest in special pipettors to gain all the advantages of low force pipetting. These ultimate tips feature a uniquely thin and flexible top that conforms to the pipet barrel unlike any other tips. They also include a non-beveled point that is so precise you can easily expel the tiniest amount of sample. And the top mounting is beveled to ensure that multichannel pipettes are easy to mount. FlexTop™ tip are made of clear 100% virgin polypropylene. Like all Labcon tips these tips are autoclavable along with their racks.