Learn More
Labcon North America Labcon Microcentrifuge Tube, 1.5 mL, Polypropylene, Amber
Catalog No. NC3995552
Encompass_Preferred
The 1.5 mL SuperSpin® Microcentrifuge Tubes in Amber offer UVSafe™ technology for protecting light-sensitive samples. Made from 100% virgin polypropylene, these tubes are designed for high-speed centrifugation and suitable for diverse lab applications in molecular biology and biochemistry.
  • Withstands centrifugation up to 40,000 x G.
  • Extra thick, durable walls.
  • Flat, pierceable caps with ergonomic 90° hinge.
  • Ultra-smooth, low retention surfaces; clear view; side writing area.
  • Autoclavable and freezable.
  • Certified endotoxin, nuclease, ATP, and heavy metals free.
  • Validated radiation sterilization.
  • USP Class VI, California Prop 65, and CE Regulation 2017/746 compliant.
  • ISO 9001:2015 quality registered; Made in USA.
Catalog No. NC3995552 Supplier Labcon North America Supplier No. 30168710009
The 1.5 mL SuperSpin® Microcentrifuge Tubes in Amber offer UVSafe™ technology for protecting light-sensitive samples. Made from 100% virgin polypropylene, these tubes are designed for high-speed centrifugation and suitable for diverse lab applications in molecular biology and biochemistry.
  • Withstands centrifugation up to 40,000 x G.
  • Extra thick, durable walls.
  • Flat, pierceable caps with ergonomic 90° hinge.
  • Ultra-smooth, low retention surfaces; clear view; side writing area.
  • Autoclavable and freezable.
  • Certified endotoxin, nuclease, ATP, and heavy metals free.
  • Validated radiation sterilization.
  • USP Class VI, California Prop 65, and CE Regulation 2017/746 compliant.
  • ISO 9001:2015 quality registered; Made in USA.