Learn More
Labcon North America Labcon Microcentrifuge Tubes, 1.5 mL, Polypropylene, Orange, 40,000 x G
Catalog No. NC3995553
Encompass_Preferred
These 1.5 mL SuperSpin® microcentrifuge tubes are made from USP Class VI polypropylene, designed for high-speed centrifugation up to 40,000 x G. Ultra-clear and durable with ergonomic, self-aligning caps, they are supplied in resealable bags for optimal sample integrity.
  • Withstands centrifugation to 40,000 x G.
  • Made from USP Class VI polypropylene.
  • Ultra-smooth, low retention interior surfaces.
  • Ergonomic caps with a 90° hinge.
  • Flat pierceable caps and a frosted writing surface.
  • Autoclavable and freezable to -80°C.
  • Endotoxin-free, RNase/DNase-free, ATP-free, and heavy metals-free.
  • Packaged in resealable medical grade bags, 5000 tubes per case.
  • Validated by radiation sterilization (SAL of 10-6).
  • Orange color for easy sample identification.
Catalog No. NC3995553 Supplier Labcon North America Supplier No. 30168730009
These 1.5 mL SuperSpin® microcentrifuge tubes are made from USP Class VI polypropylene, designed for high-speed centrifugation up to 40,000 x G. Ultra-clear and durable with ergonomic, self-aligning caps, they are supplied in resealable bags for optimal sample integrity.
  • Withstands centrifugation to 40,000 x G.
  • Made from USP Class VI polypropylene.
  • Ultra-smooth, low retention interior surfaces.
  • Ergonomic caps with a 90° hinge.
  • Flat pierceable caps and a frosted writing surface.
  • Autoclavable and freezable to -80°C.
  • Endotoxin-free, RNase/DNase-free, ATP-free, and heavy metals-free.
  • Packaged in resealable medical grade bags, 5000 tubes per case.
  • Validated by radiation sterilization (SAL of 10-6).
  • Orange color for easy sample identification.