Learn More
Labcon North America SuperClear Culture Tubes, Extra Thick Walls, Autoclavable Up to 122°C, 1000 Per Case
Catalog No. NC2121126
Encompass_Preferred
Labcon SuperClear Culture Tubes are designed for optimal laboratory use with features like extra-thick walls to enhance durability. Made from USP Class VI polypropylene resin, these 12x75 mm yellow tubes hold 5 mL and are certified free of ATP, RNase/DNase, and endotoxins for reliable results. Their round bottom design is compatible with standard rotors, and they are autoclavable up to 122°C while also standing up to freezing conditions. Packaged 1000 per case, they are ideal for cell culture applications.
This item is not returnable. View return policy
Catalog No. NC2121126 Supplier Labcon North America Supplier No. 33118020009
Labcon SuperClear Culture Tubes are designed for optimal laboratory use with features like extra-thick walls to enhance durability. Made from USP Class VI polypropylene resin, these 12x75 mm yellow tubes hold 5 mL and are certified free of ATP, RNase/DNase, and endotoxins for reliable results. Their round bottom design is compatible with standard rotors, and they are autoclavable up to 122°C while also standing up to freezing conditions. Packaged 1000 per case, they are ideal for cell culture applications.